Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC QTX125 TFA | 2989537-78-0 | 99.9% | 10 MG
SDP

Catalog No. 5000234531
Encompass_Preferred

QTX125 TFA is the trifluoroacetic acid salt of QTX125, a potent and highly selective histone deacetylase 6 (HDAC6) inhibitor with reported antitumor activity. It is supplied as a high-purity research compound for biochemical and in vivo studies.

  • Potent, selective HDAC6 inhibitor.
  • High purity: 99.9%.
  • Molecular weight: 531.44 g/mol; formula: C25H20F3N3O7.
  • Solubility: soluble in DMSO (125 mg/mL), insoluble in water.
  • Storage: store sealed at 4°C; stock solutions -80°C (6 months), -20°C (1 month).
  • Available in small research sizes including 10 MG.

Catalog No. 50-002-34531 Supplier Medchemexpress LLC Supplier No. HY120448A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

QTX125 TFA is the trifluoroacetic acid salt of QTX125, a potent and highly selective histone deacetylase 6 (HDAC6) inhibitor with reported antitumor activity. It is supplied as a high-purity research compound for biochemical and in vivo studies.

  • Potent, selective HDAC6 inhibitor.
  • High purity: 99.9%.
  • Molecular weight: 531.44 g/mol; formula: C25H20F3N3O7.
  • Solubility: soluble in DMSO (125 mg/mL), insoluble in water.
  • Storage: store sealed at 4°C; stock solutions -80°C (6 months), -20°C (1 month).
  • Available in small research sizes including 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.