Learn More
Medchemexpress LLC Qtx125 tfa | 2989537-78-0 | 99.9% | 25 MG
SDP
Catalog No. 5000234396
Encompass_Preferred
QTX125 TFA is the trifluoroacetate salt of QTX125, a potent and highly selective HDAC6 inhibitor with reported antitumor activity. It is supplied as a high-purity off-white to gray solid for biochemical and cell-based research.
  • Potent, selective inhibition of HDAC6.
  • High purity (99.9%) suitable for research applications.
  • Available in 25 MG solid packs for laboratory use.
  • Stable when stored sealed away from moisture; recommended storage at 4°C, with longer-term solvent storage at -80°C.
  • Applicable to biochemical assays and cell-based antitumor studies.
Catalog No. 50-002-34396 Supplier Medchemexpress LLC Supplier No. HY120448A25MG
QTX125 TFA is the trifluoroacetate salt of QTX125, a potent and highly selective HDAC6 inhibitor with reported antitumor activity. It is supplied as a high-purity off-white to gray solid for biochemical and cell-based research.
  • Potent, selective inhibition of HDAC6.
  • High purity (99.9%) suitable for research applications.
  • Available in 25 MG solid packs for laboratory use.
  • Stable when stored sealed away from moisture; recommended storage at 4°C, with longer-term solvent storage at -80°C.
  • Applicable to biochemical assays and cell-based antitumor studies.