Learn More
Medchemexpress LLC QTX125 trifluoroacetate | 2989537-78-0 | 99.9% | 100 MG
SDP
Catalog No. 5000232555
Encompass_Preferred
QTX125 TFA is the trifluoroacetate salt of QTX125, a potent and highly selective histone deacetylase 6 (HDAC6) inhibitor with reported antitumor activity. Supplied as an off-white to gray solid for research use, it is available in multiple pack sizes and should be stored sealed and protected from moisture.
  • Potent and selective HDAC6 inhibitor
  • High reported purity and chemical stability
  • Provided as a trifluoroacetate salt for improved handling
  • Suitable for in vitro and preclinical research applications
  • Store sealed, away from moisture; follow cold storage recommendations
Catalog No. 50-002-32555 Supplier Medchemexpress LLC Supplier No. HY120448A100MG
QTX125 TFA is the trifluoroacetate salt of QTX125, a potent and highly selective histone deacetylase 6 (HDAC6) inhibitor with reported antitumor activity. Supplied as an off-white to gray solid for research use, it is available in multiple pack sizes and should be stored sealed and protected from moisture.
  • Potent and selective HDAC6 inhibitor
  • High reported purity and chemical stability
  • Provided as a trifluoroacetate salt for improved handling
  • Suitable for in vitro and preclinical research applications
  • Store sealed, away from moisture; follow cold storage recommendations