Learn More
Alkali Scientific 0.1mL Biorad CFX/Thermo Type 96-Well PCR Plate, Low Profile, No Skirt, Clear Color, 20/pk
SDP
Supplier:  Alkali Scientific PCY9620
Encompass_Preferred
Universal Compatibility: This PCR plate is compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your lab. Easy Well Identification: High-contrast, black alphanumeric indexing facilitates quick and accurate well identification, streamlining your workflow. Optimal Heat Transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility. Sample Protection: Well ridges help reduce sample evaporation when sealing with film, foil, or strip caps, preserving the integrity of your samples. High Clarity, Ultra-Pure Material: This PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards. Purity Assurance: The plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors, and has been tested to be pyrogen-free. Nearly Identical To: Biorad item MLL9601 and additionally Thermo Scientific item AB-0700 plate. Direct substitutes.
Catalog No. 50-238-8835