Learn More
Julabo USA Inc Pair, Quad adapter M16x1 female to 4x M16x1 male with 8mm and 12mm hose barbs
SDP
Catalog No. 507534232
Encompass_Preferred
Bath Accessories, Julabo, 2 Quad distributing adapters, M16x1, with barbed fittings for tubing 8 mm or 12 mm or 1/2″ ID. Set of two circular quad manifolds with internal baffles for constant water pressure and temperature to all outlets. M16x1 screw thread fittings (which can be fitted with shut off valves), comes complete with all barbed fittings for both 8mm and 12mm/1/2″ ID tubing. For use with F1000, F500, FL1201, FL1701, FL601 and FLW1701 chillers.
Catalog No. 50-753-4232 Supplier Julabo USA Inc Supplier No. 8970520
Bath Accessories, Julabo, 2 Quad distributing adapters, M16x1, with barbed fittings for tubing 8 mm or 12 mm or 1/2″ ID. Set of two circular quad manifolds with internal baffles for constant water pressure and temperature to all outlets. M16x1 screw thread fittings (which can be fitted with shut off valves), comes complete with all barbed fittings for both 8mm and 12mm/1/2″ ID tubing. For use with F1000, F500, FL1201, FL1701, FL601 and FLW1701 chillers.