Learn More
Julabo USA Inc Pair, Quad adapter G 1 1/4"female to 4x G 1 1/4" male with 1" hose barbs
SDP
Catalog No. 507534234
Encompass_Preferred
Bath Accessories, Julabo, 2 Quad distributing adapters, G 1 1/4″ female, with barbed fittings for tubing 1″ ID. Set of two circular quad manifolds with internal baffles for constant water pressure and temperature to all outlets. G 1¼” female thread fittings (which can be fitted with shut off valves), comes complete with all barbed fittings for 1” ID tubing. For use with FL2506, FLW2506, FL4006, FLW4006, FL7006, FLW7006, FL11006, FLW11006, FL20006, FLW20006 chillers.
Catalog No. 50-753-4234 Supplier Julabo USA Inc Supplier No. 8970524
Bath Accessories, Julabo, 2 Quad distributing adapters, G 1 1/4″ female, with barbed fittings for tubing 1″ ID. Set of two circular quad manifolds with internal baffles for constant water pressure and temperature to all outlets. G 1¼” female thread fittings (which can be fitted with shut off valves), comes complete with all barbed fittings for 1” ID tubing. For use with FL2506, FLW2506, FL4006, FLW4006, FL7006, FLW7006, FL11006, FLW11006, FL20006, FLW20006 chillers.