Learn More
Julabo USA Inc Pair, Quad adapter G 3/4"female to 4x G 3/4" male with 3/4" hose barbs
SDP
Catalog No. 507534233
Encompass_Preferred
Bath Accessories, Julabo, 2 Quad distributing adapters, G3/4″ female, with barbed fittings for tubing 3/4″ ID. Set of two circular quad manifolds with internal baffles for constant water pressure and temperature to all outlets. G 3/4″ female thread fittings (which can be fitted with shut off valves), comes complete with all barbed fittings for ¾” ID tubing. For use with FL1203, FL1703, FL2503, FL4003, FLW1703, FLW2503, FLW4003 chillers.
Catalog No. 50-753-4233 Supplier Julabo USA Inc Supplier No. 8970522
Bath Accessories, Julabo, 2 Quad distributing adapters, G3/4″ female, with barbed fittings for tubing 3/4″ ID. Set of two circular quad manifolds with internal baffles for constant water pressure and temperature to all outlets. G 3/4″ female thread fittings (which can be fitted with shut off valves), comes complete with all barbed fittings for ¾” ID tubing. For use with FL1203, FL1703, FL2503, FL4003, FLW1703, FLW2503, FLW4003 chillers.