Learn More
Bellco Glass, Inc. Quad Additon Assembly , 45mm Universal
SDP
Catalog No. 501944751
Encompass_Preferred
Quad Addition Assembly For GL45 sidearms. Manufactured from 316l stainless steel with electropolish finish ra 32 or better outside only. Used for gl45 sidearms. Includes polypropylene blue open top screw cap and white silicone liner. Individual ports for liquid additions or redundant vent overlay. Vertical Side Arm vessels for acid, base, antifoam, media & other additions. MADE IN THE USA
Catalog No. 50-194-4751 Supplier Bellco Glass, Inc. Supplier No. 196404125
Quad Addition Assembly For GL45 sidearms. Manufactured from 316l stainless steel with electropolish finish ra 32 or better outside only. Used for gl45 sidearms. Includes polypropylene blue open top screw cap and white silicone liner. Individual ports for liquid additions or redundant vent overlay. Vertical Side Arm vessels for acid, base, antifoam, media & other additions. MADE IN THE USA