Learn More
Electron Microscopy Sciences Quadruple Geological Thin Section Holder, Hitachi M4
SDP
Catalog No. 501901648
Encompass
This thin section holder made from aluminum with copper clips and brass screws. The unit measures 107x58x8 mm and it holds four standard geological thin sections or slides of 47 x 27 mm. The holder has convenient tweezer inserts which allow for easy loading. Clips are provided for secure holding.
Type: Hitachi M4
Catalog No. 50-190-1648 Supplier Electron Microscopy Sciences Supplier No. 7594119H
This thin section holder made from aluminum with copper clips and brass screws. The unit measures 107x58x8 mm and it holds four standard geological thin sections or slides of 47 x 27 mm. The holder has convenient tweezer inserts which allow for easy loading. Clips are provided for secure holding.
Type: Hitachi M4