Learn More
Waters Corp WATERS CORPORATION QC Reference Material, Quad LCMS, Eight-Component Mix, 1 mL
Catalog No. 50727072 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Quad LCMS QC Reference Material is designed to benchmark and assess the performance of UPLC systems, including those equipped with single or triple quad mass spectrometers. It contains a 1 mL eight-component mix of various compounds in an Acetonitrile/water solution with formic acid.
  • Intended for verifying the performance of UPLC systems, especially those with single or triple quad mass spectrometers.
  • Contains a comprehensive mix of eight compounds: 100 ug/mL Acetaminophen, 50 ug/mL Sulfaguanidine, 10 ug/mL Sulfadimethoxine, 25 ug/mL Val,-Tyr,-Val, 6 ug/mL Verapamil, 6 ug/mL Terfenadine, 25 ug/mL Leucine Enkephalin, and 18 ug/mL Reserpine.
  • Compounds are dissolved in 19/81 Acetonitrile/water with 0.008% formic acid.
Catalog No. 50-727-072 Supplier Waters Corp Supplier No. 186007362
This Quad LCMS QC Reference Material is designed to benchmark and assess the performance of UPLC systems, including those equipped with single or triple quad mass spectrometers. It contains a 1 mL eight-component mix of various compounds in an Acetonitrile/water solution with formic acid.
  • Intended for verifying the performance of UPLC systems, especially those with single or triple quad mass spectrometers.
  • Contains a comprehensive mix of eight compounds: 100 ug/mL Acetaminophen, 50 ug/mL Sulfaguanidine, 10 ug/mL Sulfadimethoxine, 25 ug/mL Val,-Tyr,-Val, 6 ug/mL Verapamil, 6 ug/mL Terfenadine, 25 ug/mL Leucine Enkephalin, and 18 ug/mL Reserpine.
  • Compounds are dissolved in 19/81 Acetonitrile/water with 0.008% formic acid.