Learn More
Braintree Scientific Quad Mouse Irradiation Base Plate for head exposure, 1.375""
SDP
Catalog No. 501955074
Encompass
The shields are not suitable for 360 degree irradiation (typical of a Cs source), and they are limited for use in units that produce energies less than 350kVp (X-Ray). The fixtures are to be used with fixed beam X-ray sources projecting vertically and do not protect the exit side of the irradiated specimen. A shield designed for use with a Cs source typically requires 4 times the lead thickness. These shields are " (6.35mm) lead, and the attenuation factor depends on the kVp energy they are used with. Attempts to use our shields with a Cs source will result in inadequate shielding and overexposure to protected areas of an animal. The shields are regularly used in the Orthovoltage energy range (<320 kVp) and provide about a 100X attenuation. See Attenuation Curves for a better evaluation. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5074 Supplier Braintree Scientific Supplier No. MHS2HQBPS
The shields are not suitable for 360 degree irradiation (typical of a Cs source), and they are limited for use in units that produce energies less than 350kVp (X-Ray). The fixtures are to be used with fixed beam X-ray sources projecting vertically and do not protect the exit side of the irradiated specimen. A shield designed for use with a Cs source typically requires 4 times the lead thickness. These shields are " (6.35mm) lead, and the attenuation factor depends on the kVp energy they are used with. Attempts to use our shields with a Cs source will result in inadequate shielding and overexposure to protected areas of an animal. The shields are regularly used in the Orthovoltage energy range (<320 kVp) and provide about a 100X attenuation. See Attenuation Curves for a better evaluation. **Sold for research purposes only and not for human use**