Learn More
Chemglass Life Sciences Quad Port Adapter, (4) 1/8" Angled Hose Barbs, M18
SDP
Catalog No. NC3723199
Encompass
The Quad inlet assembly fits into an 18mm head plate port.The assembly incorporates four separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.M18 threaded adapter has (4) 1/8 (3. 2mm) hose barb tubing ports for connection to flexible tubing.Inside the reactor, the 1/8 tubes are 5/8, 3/4, 1 and 1-1/8 long.
Catalog No. NC3723199 Supplier Chemglass Life Sciences Supplier No. CLS1380411
The Quad inlet assembly fits into an 18mm head plate port.The assembly incorporates four separate pipes into one assembly for efficient use of space.Each port can be used individually for reagent addition.M18 threaded adapter has (4) 1/8 (3. 2mm) hose barb tubing ports for connection to flexible tubing.Inside the reactor, the 1/8 tubes are 5/8, 3/4, 1 and 1-1/8 long.