Learn More
Chemglass Life Sciences Quad Port Adapter, M18 Thread, for Bio Reactors
SDP
Catalog No. NC2131792
Encompass
The Quad inlet assembly fits into an 18mm head plate port.The assembly incorporates four separate pipes into one assembly for efficient use of space.Each port can be used indivually for reagent addition.M18 threaded adapter has (4) 1/8" (3.2mm) tubing ports for connection to flexible tubing.Inside the reactor, the 1/8" tubes are 5/8", 3/4", 1" and 1-1/8" long.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
This item is not returnable. View return policy
Catalog No. NC2131792 Supplier Chemglass Life Sciences Supplier No. CLS1380409
The Quad inlet assembly fits into an 18mm head plate port.The assembly incorporates four separate pipes into one assembly for efficient use of space.Each port can be used indivually for reagent addition.M18 threaded adapter has (4) 1/8" (3.2mm) tubing ports for connection to flexible tubing.Inside the reactor, the 1/8" tubes are 5/8", 3/4", 1" and 1-1/8" long.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.