Learn More
MSC Bird-X Ultrasonic Bird Repeller, 6,500 sq ft Coverage, 4 Speakers
Catalog No. 502836217
Encompass
The QuadBlaster QB-4 is an ultrasonic electronic pest repeller designed for indoor and semi-enclosed areas. It utilizes four speakers to emit high-frequency sound waves, effectively deterring birds and other small pests from areas up to 6,500 sq ft.
  • Utilizes ultrasonic technology, silent to humans
  • Features four speakers for broad sound distribution
  • Deters various pests including birds, bats, rats, and mice
  • Provides coverage for up to 6,500 sq ft
  • Suitable for indoor and semi-enclosed environments
  • Designed for simple mounting
  • Helps protect buildings and stored equipment from damage
  • UL and CE listed for compliance
  • Operates within a 20-30 kHz frequency range
  • Low energy consumption of less than 10 watts
Catalog No. 50-283-6217 Supplier MSC Supplier No. 72510837
The QuadBlaster QB-4 is an ultrasonic electronic pest repeller designed for indoor and semi-enclosed areas. It utilizes four speakers to emit high-frequency sound waves, effectively deterring birds and other small pests from areas up to 6,500 sq ft.
  • Utilizes ultrasonic technology, silent to humans
  • Features four speakers for broad sound distribution
  • Deters various pests including birds, bats, rats, and mice
  • Provides coverage for up to 6,500 sq ft
  • Suitable for indoor and semi-enclosed environments
  • Designed for simple mounting
  • Helps protect buildings and stored equipment from damage
  • UL and CE listed for compliance
  • Operates within a 20-30 kHz frequency range
  • Low energy consumption of less than 10 watts