Learn More
Electron Microscopy Sciences QuadmagGenie, 50hz, Australian Plug, 230 V
SDP
Catalog No. 50949831
Encompass_Preferred
The powerful multi-position QuadMag Genie magnetic stirrer provides vigorous stirring for up to 4 vessels at a time. Designed for general laboratory purposes, it features a powerful magnet and motor that offer exceptional magnetic force capable of stirring up to 2L for each position. Stir bars are pulled into position and stay coupled even under the most demanding stirring conditions.
Rugged ABS/Polycarbonate housing and the famous Vortex-Genie heavy-duty motor provide years of reliable service.
Easy-to-use controls allow users to easily change the desired speed.
Spill-resistant housing channels fluids away from the internal components.
Can be used in cold rooms and incubators.
Non-skid feet.
Catalog No. 50-949-831 Supplier Electron Microscopy Sciences Supplier No. 72402AU
The powerful multi-position QuadMag Genie magnetic stirrer provides vigorous stirring for up to 4 vessels at a time. Designed for general laboratory purposes, it features a powerful magnet and motor that offer exceptional magnetic force capable of stirring up to 2L for each position. Stir bars are pulled into position and stay coupled even under the most demanding stirring conditions.
Rugged ABS/Polycarbonate housing and the famous Vortex-Genie heavy-duty motor provide years of reliable service.
Easy-to-use controls allow users to easily change the desired speed.
Spill-resistant housing channels fluids away from the internal components.
Can be used in cold rooms and incubators.
Non-skid feet.