Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuadraPure™ IMDAZ, 100-400 micron, metal scavenger, Thermo Scientific™

Catalog No. AA4613306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4613306 5 g
AA4613314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4613306 Supplier Thermo Scientific Chemicals Supplier No. 4613306

Metal Scavenger: Pd, Ru, Os, Co, Ni, Rh, V, Fe, Cu, Sn

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Chemical Name or Material QuadraPure™ IMDAZ metal scavenger
Physical Form Beads
MDL Number MFCD07785599
UN Number UN3082
Synonym QuadraPure™ Imidazol-1-yl propylaminobut-2-enoate ester
Quantity 5 g
Particle Size 100 to 400 Micron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.