Learn More
Chemglass Life Sciences QuadRocker Rocker, 115V, 60Hz, Benchmark
SDP
Catalog No. 5000083282
Encompass_Preferred
QuadRocker features a unique, tiered attachment system that accepts up to four platforms. Unlike traditional single platform rockers, the QuadRocker includes a base level as well as three additional stacking platforms, providing a total work-surface of over 750 sq. in. For most gel blotting, washing and staining applications, the rocker can be installed with all four rocking platforms. For applications requiring larger vessels, the quantity and height of the platforms can easily be customized to provide a maximum clearance of up to 8-1/4 in (21cm). Digital speed and timer control, as well as user-adjustable tilt angle. Compact footprint. Accepts up to four tiers. User-adjustable platform configuration. Adjustable rocking speed and tilt angle. Cold room and incubator safe. Includes four large platforms.
Catalog No. 50-000-83282 Supplier Chemglass Life Sciences Supplier No. CLS402501
QuadRocker features a unique, tiered attachment system that accepts up to four platforms. Unlike traditional single platform rockers, the QuadRocker includes a base level as well as three additional stacking platforms, providing a total work-surface of over 750 sq. in. For most gel blotting, washing and staining applications, the rocker can be installed with all four rocking platforms. For applications requiring larger vessels, the quantity and height of the platforms can easily be customized to provide a maximum clearance of up to 8-1/4 in (21cm). Digital speed and timer control, as well as user-adjustable tilt angle. Compact footprint. Accepts up to four tiers. User-adjustable platform configuration. Adjustable rocking speed and tilt angle. Cold room and incubator safe. Includes four large platforms.