Learn More
Apexbio Technology LLC Quadrol 102-60-3 1g
SDP
Catalog No. 5000089709
Encompass_Preferred
Quadrol is an immunostimulatory molecule designed to modulate immune responses by influencing cytokine signaling cascades and chemotaxis of immune cells Quadrol exerts its biological activity primarily by regulating immune cell recruitment and altering inflammatory mediator dynamics Experimental studies employing in vitro cellular models and preclinical animal wound injury models have utilized Quadrol to investigate its effects on immune cell migration inflammatory processes and subsequent tissue regeneration Based on these pharmacological properties Quadrol holds research potential in tissue repair and wound healing applications
Catalog No. 50-000-89709 Supplier Apexbio Technology LLC Supplier No. C67711000
Quadrol is an immunostimulatory molecule designed to modulate immune responses by influencing cytokine signaling cascades and chemotaxis of immune cells Quadrol exerts its biological activity primarily by regulating immune cell recruitment and altering inflammatory mediator dynamics Experimental studies employing in vitro cellular models and preclinical animal wound injury models have utilized Quadrol to investigate its effects on immune cell migration inflammatory processes and subsequent tissue regeneration Based on these pharmacological properties Quadrol holds research potential in tissue repair and wound healing applications