Learn More
Medchemexpress LLC Quadrol (N,N,N′,N′-Tetrakis(2-hydroxypropyl)ethylenediamine) | 102-60-3 | 99.9% | 500 MG
SDP
Catalog No. 5000409998
Encompass_Preferred
Quadrol is an immunostimulant that has been considered as a potentially useful active molecule for accelerating wound healing. It can form a complex with Ca2+, mediate the entry of Ca2+ into macrophages, and stimulate macrophage activation.
  • Immunostimulant.
  • Potentially useful for accelerating wound healing.
  • Forms a complex with Ca2+.
  • Mediates the entry of Ca2+ into macrophages.
  • Stimulates macrophage activation.
  • In vitro, concentrations of 1 mM and 4 mM enhanced macrophage spreading and phagocytic activity.
  • In vivo, it promoted wound collagen formation in normal and diabetic mice.
Catalog No. 50-004-09998 Supplier Medchemexpress LLC Supplier No. HYB2149500MG
Quadrol is an immunostimulant that has been considered as a potentially useful active molecule for accelerating wound healing. It can form a complex with Ca2+, mediate the entry of Ca2+ into macrophages, and stimulate macrophage activation.
  • Immunostimulant.
  • Potentially useful for accelerating wound healing.
  • Forms a complex with Ca2+.
  • Mediates the entry of Ca2+ into macrophages.
  • Stimulates macrophage activation.
  • In vitro, concentrations of 1 mM and 4 mM enhanced macrophage spreading and phagocytic activity.
  • In vivo, it promoted wound collagen formation in normal and diabetic mice.