Learn More
Ready America™ QuakeHOLD!™ Flex Mount Fastener for 200 pounds
SDP
Catalog No. 11900053
Change view
Click to view available options
Includes: Two 12 in. Bio Straps with Carabiners, Two 2 x 3 in. Thumb Lock Fasteners With 3M(R) VHB Adhesive, Two Surface Prep Pads, Instruction Sheet
Catalog No.
11-900-053 Includes: Two 12 in. Bio Straps with Carabiners, Two 2 x 3 in. Thumb Lock Fasteners With 3M(R) VHB Adhesive, Two Surface Prep Pads, Instruction Sheet
1 options
Catalog No. 11-900-053 Supplier Ready America™ Supplier No. RDFMT62
Add to Cart
Add to Cart
Ready America™ QuakeHOLD!™ Flex Mount Fastener for 200 pounds is suitable for bracing flat surface equipment by attaching to your unit with the opposing end secured to the flexmount bar.
  • Secures flat surface equipment
  • Used in conjunction with Flex Mount Bar
  • Ideal for IBC Compliance
  • Features 3M™ VHB adhesive
  • 100 lb weight capacity