Learn More
Ready America™ QuakeHOLD!™ Flex Mount Fastener
SDP
Catalog No. 11900055
Change view
Click to view available options
Includes: 12 in. Bio Strap with Buckle and Carabiner, Instruction Sheet
Catalog No.
11-900-055 Includes: 12 in. Bio Strap with Buckle and Carabiner, Instruction Sheet
1 options
Catalog No. 11-900-055 Supplier Ready America™ Supplier No. RDFMMIC1
  • $42.25
    $26.65 / Each of 1
    Save $15.60
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Ready America™ QuakeHOLD!™ Flex Mount Fastener is suitable for bracing flat surface equipment by attaching to your unit with the opposing end secured to the flexmount bar.
  • Secures flat surface equipment
  • Used in conjunction with Flex Mount Bar
  • Ideal for IBC Compliance
  • Features 3M™ VHB adhesive
  • 100 lb weight capacity