Learn More
Ready America™ QuakeHOLD!™ Flex Mount Silver Fastener
SDP
Catalog No. 11900056
Change view
Click to view available options
Includes: Two 12 in. Cable with Carabiners, Two 2 x 3 in. Stainless Steel 316 Plates with 3M(R) VHB Adhesive, Two Surface Prep Pads, Instruction Sheet
Catalog No.
11-900-056 Includes: Two 12 in. Cable with Carabiners, Two 2 x 3 in. Stainless Steel 316 Plates with 3M(R) VHB Adhesive, Two Surface Prep Pads, Instruction Sheet
1 options
Catalog No. 11-900-056 Supplier Ready America™ Supplier No. RDFM23C2
  • $168.00
    $130.65 / Each of 1
    Save $37.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Ready America™ QuakeHOLD!™ Flex Mount Silver Fastener is suitable for bracing flat surface equipment by attaching to your unit with the opposing end secured to the flexmount bar.
  • Secures flat surface equipment
  • Used in conjunction with Flex Mount Bar
  • Ideal for IBC Compliance
  • Features 3M™ VHB adhesive
  • 200 lb weight capacity