Learn More
Ready America™ QuakeHOLD!™ Large Floor Equipment Bio Coated Strut Strap
SDP
Catalog No. 11900436
Change view
Click to view available options
Color: Black
Catalog No.
11-900-436 Color: Black
1 options
Catalog No. 11-900-436 Supplier Ready America™ Supplier No. RWLFEB
  • $419.00
    $265.65 / Each of 1
    Save $153.35
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Ready America™ QuakeHOLD!™ Large Floor Equipment Bio Coated Strut Strap for securing freezers, refrigerators, shakers, incubators and other large and standing equipment.
Includes the following:
  • 2 2 x 32 in. Coated Straps With Strut Mounting Bracket
  • 2 4 x 6 in. Aluminum Plates With 3M™ Vhb Adhesive
  • 2 2.5 x 3.5 in. Plate
  • 4 Wing Nuts
  • 4 0.25 in. Machine Screws
  • 4 0.25 in. Bolts
  • 8 0.2 x 0.75 in. Washers
  • 2 0.2 in. Strut-Washer Nuts
  • 2 0.2 x 1 in. Bolts
  • 2 0.2 x 1 in. Flat Washers
  • 2 Surface Prep Pads