Learn More
BURKLE INC QualiSampler set PTFE/FEP, compl. with carrying case
Catalog No. 5000025600
Encompass_Preferred
The handy QualiSampler set, complete with a carrying case, combines the LiquiSampler and ViscoSampler—both made from PTFE/FEP—along with practical accessories. It is perfectly suited for sampling liquids and viscous substances.
Catalog No. 50-000-25600 Supplier BURKLE INC Supplier No. 53301001
The handy QualiSampler set, complete with a carrying case, combines the LiquiSampler and ViscoSampler—both made from PTFE/FEP—along with practical accessories. It is perfectly suited for sampling liquids and viscous substances.