Learn More
Dentec Safety™ Qualitative Fit Test Ampules
SDP
Catalog No. 19222539 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
19-222-539 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 19-222-539 Supplier Dentec Safety™ Supplier No. 706200
Add to Cart
Add to Cart
This item is not returnable. View return policy
Banana oil (isoamyl acetate) has a strong, recognizable odor to quickly indicate the lack of proper respirator fit.
  • Qualitative fit test ampules
  • Banana Oil (IAA)
  • 10/Pk.
Specifications
Product Type Respirator Accessory
For Use With (Equipment) Masks
Includes Respirator accessories
Model 706200
Quantity 10/Pk.