Learn More
BTNX Inc Rapid Response™ Qualitative Urine Toxicology Control – FYL
Catalog No. NC2023915
Encompass_Preferred
The Rapid Response™ Qualitative Urine Toxicology Control is packaged in amber vials (borosilicate glass) with silicone septas. The matrix is synthetic urine. This product lasts until the expiry date listed on the bottle, or 31 days after opening. For forensic use only.For use with Rapid Response™ Fentanyl Test Strip (Urine) (Cat. #FYL-1S48-100).
Catalog No. NC2023915 Supplier BTNX Inc Supplier No. RR1VPNFYL.48
The Rapid Response™ Qualitative Urine Toxicology Control is packaged in amber vials (borosilicate glass) with silicone septas. The matrix is synthetic urine. This product lasts until the expiry date listed on the bottle, or 31 days after opening. For forensic use only.For use with Rapid Response™ Fentanyl Test Strip (Urine) (Cat. #FYL-1S48-100).