Learn More
Perkin Elmer US LLC Quality Control Standard 1B, 10% HNO3, 125 mL
Catalog No. 502104514
Encompass_Preferred
Details Concentration Element 1,000 µg/mL As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Se, Sr, Ti, Tl, V, Zn,
Catalog No. 50-210-4514 Supplier Perkin Elmer US LLC Supplier No. N9304131
Details Concentration Element 1,000 µg/mL As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Se, Sr, Ti, Tl, V, Zn,