Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Control Standard 7A, Ricca Chemical
SDP

Catalog No. RQC7A100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RQC7A-100 100 mL
RQC7A-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RQC7A-100 Supplier Ricca Chemical Company Supplier No. RQC7A100
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Quality Control Standard 7A
Packaging Amber Poly Bottle
CAS 7697-37-2, 7732-18-5, 7757-79-1, 7784-27-2, 10043-35-3
CAS Min % 90.0, 7.64, 0.28, 0.23, 0.11, 0.05
CAS Max % 93.0, 7.95, 0.29, 0.24, 0.12, 0.05
Odor Odorless
Color Colorless
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.