Learn More
Waters Corp QuanRecovery Plates, 700 μL, MaxPeak HPS, 96-Well
Catalog No. NC1723860 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These plates are designed for LC-MS bioanalysis, engineered to reduce peptide and protein losses due to ionic interactions and non-specific binding. This design leads to higher recovery, sensitivity, and repeatability in analytical results by minimizing analyte/surface interactions.
  • Reduce peptide and protein losses due to ionic interactions and non-specific binding.
  • LC-MS autosampler ready.
  • Low residual volumes for efficient utilization of low sample volumes.
  • MaxPeak High Performance Surfaces minimize analyte/surface interactions, increasing analyte recovery, sensitivity, and reproducibility.
Catalog No. NC1723860 Supplier Waters Corp Supplier No. 186009184
These plates are designed for LC-MS bioanalysis, engineered to reduce peptide and protein losses due to ionic interactions and non-specific binding. This design leads to higher recovery, sensitivity, and repeatability in analytical results by minimizing analyte/surface interactions.
  • Reduce peptide and protein losses due to ionic interactions and non-specific binding.
  • LC-MS autosampler ready.
  • Low residual volumes for efficient utilization of low sample volumes.
  • MaxPeak High Performance Surfaces minimize analyte/surface interactions, increasing analyte recovery, sensitivity, and reproducibility.