Learn More
Waters Corp QuanRecovery with MaxPeak, 700 µL Plates, 5/pk
Catalog No. NC1705395 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed by LC-MS bioanalysts for LC-MS bioanalysts, QuanRecovery Plates with MaxPeak HPS reduce peptide and protein losses due to ionic interactions and non specific binding. A reduction in these losses lead to higher recovery, sensitivity and repeatability in analytical results. These 700 µL 96 well plates are LC-MS autosampler ready, with low residual volumes to fully utilize low sample volumes. MaxPeak High Performance Surfaces are new and innovative technologies designed to increase analyte recovery, sensitivity and reproducibility by minimizing analyte/surface interactions that can lead to sample losses.
Catalog No. NC1705395 Supplier Waters Corp Supplier No. 186009185
Designed by LC-MS bioanalysts for LC-MS bioanalysts, QuanRecovery Plates with MaxPeak HPS reduce peptide and protein losses due to ionic interactions and non specific binding. A reduction in these losses lead to higher recovery, sensitivity and repeatability in analytical results. These 700 µL 96 well plates are LC-MS autosampler ready, with low residual volumes to fully utilize low sample volumes. MaxPeak High Performance Surfaces are new and innovative technologies designed to increase analyte recovery, sensitivity and reproducibility by minimizing analyte/surface interactions that can lead to sample losses.