Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Quant-iT™ PicoGreen ReadyPlate

Catalog No. P32705
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96-Well ReadyPlate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P32705 1 x 96-Well ReadyPlate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P32705 Supplier Invitrogen™ Supplier No. P32705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Quant-iT PicoGreen ReadyPlate is a preloaded 96-well plate that offers a rapid, ultra-sensitive, and selective method for double-stranded DNA quantification. The plate is prefilled with buffer and PicoGreen reagent, which exhibits bright green fluorescence upon binding to DNA.

The streamlined assay protocol requires users to simply hydrate the wells, add the provided standards and user samples, and incubate for two minutes prior to fluorescent measurement. This format allows users to quantify 0.2–200 ng of DNA (0.002–2 ng/μL when using 100 μL samples).

The Quant-iT PicoGreen ReadyPlate is ideal for quantitating small amounts of DNA for many applications, including synthesizing cDNA for library production, quantifying DNA libraries for next-generation sequencing, and purifying DNA fragments for subcloning.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Format Kit
Label or Dye Other Labels or Dyes
Final Product Type DNA
Sample Type DNA
Labeling Target DNA
Product Type Pre-filled Microplate
Quantity 1 x 96-Well ReadyPlate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.