Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Quant-iT™ RiboGreen ReadyPlate

Catalog No. R32705
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96-Well ReadyPlate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R32705 1 x 96-Well ReadyPlate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R32705 Supplier Invitrogen™ Supplier No. R32705
Add to Cart
Add to Cart

The Quant-iT RiboGreen ReadyPlate is a preloaded 96-well plate that offers a rapid, simplified, and highly sensitive method for RNA quantification. The ReadyPlate is prefilled with buffer and RiboGreen reagent, which exhibits bright green fluorescence upon binding to RNA or DNA.

The streamlined assay format, along with prediluted standards, enables users to get to their results faster. Here, the user needs to simply hydrate the wells, add the provided standards and user samples, and incubate for two minutes prior to fluorescent measurement. This format allows users to quantify 1–200 ng of RNA (0.01–2 ng/μL when using 100 μL samples).

The Quant-iT RiboGreen ReadyPlate is ideal for measuring yields of RNA preparations for microarray experiments, reverse transcription PCR (RT-PCR), differential display PCR, northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparations, and high-throughput automated workflows.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Format Kit
Label or Dye Other Labels or Dyes
Final Product Type RNA (Labeled)
Sample Type RNA
Labeling Target RNA (General)
Product Type Pre-filled Microplate
Quantity 1 x 96-Well ReadyPlate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.