Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Quant-iT™ RNA Assay Kits and Quant-iT RNA HS Reagent

Catalog No. q33225
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 mL
1,000 assays
1000 assays
Assay:
RNA Quantitation
RNA Quantitation, BR
RNA Quantitation, HS
RNA Quantitation, ext range
Quantitation Range:
20 to 1000 ng
200 to 10,000 ng
5 to 100 ng
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Assay Quantitation Range
Q33225 1000 assays RNA Quantitation, ext range 200 to 10,000 ng
Q33140 1 kit RNA Quantitation 5 to 100 ng
Q10213 1,000 assays RNA Quantitation, BR 20 to 1000 ng
Q32884 1 mL RNA Quantitation, HS 5 to 100 ng
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. Q33225 Supplier Invitrogen™ Supplier No. Q33225
Only null left
Add to Cart
Edge
Add to Cart

Achieve highly selective and accurate quantification of high-abundance RNA samples with the fluorescent Quant-iT RNA Reagent and Assay Kits. These kits are highly selective for RNA over double-stranded DNA, and provide sufficient material for at least 2000 assays in a 96-well microplate format.

Enable easy and accurate RNA quantitation with the Quant-iT RNA Assay Kit, which is also available in broad range (BR) and extended range (XR) configurations, as well as a stand-alone high sensitivity (HS) reagent. These Quant-iT RNA quantification kits are highly selective for RNA over dsDNA and can be used to generate fluorescence-based linear detection ranges of RNA.

Quant-iT RNA Reagent and Assay Kit
The Quant-iT RNA Reagent and Assay Kit makes RNA quantification easy and accurate. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted RNA standards. The assay is highly selective for RNA over double-stranded DNA, and provides a dynamic range of 5-100 ng of RNA.

Quant-iT RNA BR (Broad Range) Assay Kit
The Quant-iT RNA BR (Broad Range) Assay Kit provides an accurate and selective method for quantification of high-abundance RNA samples. While the original Quant-iT RNA kit has great sensitivity, the Quant-iT RNA Broad Range kit is ideal for microarray samples where the concentration of RNA is higher. The assay is highly selective for RNA over double-stranded DNA, and provides a dynamic range of 20-1000 ng of RNA.

Quant-iT RNA XR (Extended Range) Assay Kit
The Quant-iT RNA XR (Extended Range) Assay Kit provides an accurate and selective method for the quantification of high-abundance RNA samples. The assay is highly selective for RNA and does not quantitate DNA, protein, or free nucleotides. The assay kit is designed to be accurate for initial RNA sample concentrations of 10 ng/μL to 10,000 ng/μL depending on sample volume, providing a linear detection range of 200–10,000 ng.

For each assay, simply dilute the reagent using the buffer provided, add the sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a fluorescence-based microplate reader. Common contaminants, such as salts, free nucleotides, solvents, detergents, and protein, are well tolerated.

TRUSTED_SUSTAINABILITY

Specifications

Assay RNA Quantitation, ext range
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Quantitation Range 200 to 10,000 ng
Product Line Quant-iT
Quantity 1000 assays
Shipping Condition Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.