Learn More
QA Bio Inc N-acetylneuraminic acid quantitative standard
Catalog No. 502236578
Encompass
The N-acetylneuraminic acid (NANA or Neu5Ac) standard is a quantitative standard of NIST-F and USP traceable Neu5Ac monosaccharide. • Part numbers: CM-NEU-AC-01 (1 nmols) / CM-NEUAC-100 (100 nmols). The N-glycolylneuraminic acid quantitative standard (Neu5Gc or NGNA) is also available.
This item is not returnable. View return policy
Catalog No. 50-223-6578 Supplier QA Bio Inc Supplier No. CMNEUAC01
The N-acetylneuraminic acid (NANA or Neu5Ac) standard is a quantitative standard of NIST-F and USP traceable Neu5Ac monosaccharide. • Part numbers: CM-NEU-AC-01 (1 nmols) / CM-NEUAC-100 (100 nmols). The N-glycolylneuraminic acid quantitative standard (Neu5Gc or NGNA) is also available.