Learn More
QA Bio Inc N-glycolyneuraminic acid quantitative standard
Catalog No. 502236579
Encompass
The N-glycoylneuraminic acid (NGNA or Neu5Gc) standard is a quantitative standard of NIST-F and USP traceable Neu5Gc monosaccharide. • Part numbers: CM-NEU-GC-01 (1 nmols) / CM-NEUGC-100 (100 nmols). The N-acetlylneuraminic acid quantitative standard (Neu5Ac or NANA) is also available.
This item is not returnable. View return policy
Catalog No. 50-223-6579 Supplier QA Bio Inc Supplier No. CMNEUGC01
The N-glycoylneuraminic acid (NGNA or Neu5Gc) standard is a quantitative standard of NIST-F and USP traceable Neu5Gc monosaccharide. • Part numbers: CM-NEU-GC-01 (1 nmols) / CM-NEUGC-100 (100 nmols). The N-acetlylneuraminic acid quantitative standard (Neu5Ac or NANA) is also available.