Learn More
Electron Microscopy Sciences R 1.2/1.3 Holey Carbon Films on Grids, Grid: Copper. Mesh: 300., 25/PK
SDP
Catalog No. 502091427
Encompass_Preferred
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. A foil with ~1.2 µm circular holes and a spacing of ~1.3 µm between the holes. Smaller holes are designed for magnifications of 50,000x or higher. Grid: Copper. Mesh: 300 .
Catalog No. 50-209-1427 Supplier Electron Microscopy Sciences Supplier No. Q325CR1.32NM
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. A foil with ~1.2 µm circular holes and a spacing of ~1.3 µm between the holes. Smaller holes are designed for magnifications of 50,000x or higher. Grid: Copper. Mesh: 300 .