Learn More
Electron Microscopy Sciences Perforated support foil with pre-defined holes has advantages in electron microscopy compared with conventional holey film. 2 µm holes with 3 µm period. Magnifications between 30,000x and 40,000x.
SDP
Catalog No. NC3641008
Encompass_Preferred
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 2 µm. Space between holes is 3 µm. Center to center is 3 µm. Types with 2 µm holes are recommended for magnifications between 30.000x and 40.000x.
Catalog No. NC3641008 Supplier Electron Microscopy Sciences Supplier No. Q410AR.1
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 2 µm. Space between holes is 3 µm. Center to center is 3 µm. Types with 2 µm holes are recommended for magnifications between 30.000x and 40.000x.