Learn More
Invivogen QUANTI-Blue™ Solution, Alkaline phosphatase detection medium - Liquid form, 5 ml
SDP
Catalog No. NC1647037
Encompass
QUANTI-Blue™ is a colorimetric enzyme assay developed to determine any alkaline phosphatase (AP) activity in a biological sample, such as cell culture supernatant. QUANTI-Blue™ Solution is a liquid formulation (100X) of QUANTI-Blue™ Powder (1X) offering a convenient, easy and rapid means to detect and quantify SEAP and AP activity.
Catalog No. NC1647037 Supplier Invivogen Supplier No. REPQBS
QUANTI-Blue™ is a colorimetric enzyme assay developed to determine any alkaline phosphatase (AP) activity in a biological sample, such as cell culture supernatant. QUANTI-Blue™ Solution is a liquid formulation (100X) of QUANTI-Blue™ Powder (1X) offering a convenient, easy and rapid means to detect and quantify SEAP and AP activity.