Learn More
Invivogen Alkaline phosphatase detection medium - Liquid form, 20 mL (100X)
SDP
Catalog No. NC1821819
Encompass
QUANTI-Blue Solution is a colorimetric enzyme assay developed to determine any alkaline phosphatase (AP) activity in a biological sample, such as cell culture supernatant. It has been optimized for use in 96-well plates (standard procedure) and in 1536-well plates (high-throughput screening procedure).
Catalog No. NC1821819 Supplier Invivogen Supplier No. REPQBS3
QUANTI-Blue Solution is a colorimetric enzyme assay developed to determine any alkaline phosphatase (AP) activity in a biological sample, such as cell culture supernatant. It has been optimized for use in 96-well plates (standard procedure) and in 1536-well plates (high-throughput screening procedure).