Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Quanti-Cult Plus™ Candida albicans ATCC™ 10231™

Catalog No. R4711503 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4711503 100 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4711503 Supplier Thermo Scientific™ Supplier No. R4711503
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.

Thermo Scientific™ Quanti-Cult Plus™ Candida albicans ATCC™ 10231™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • Ready for rehydration
  • For use in quality control procedures

Ordering Alerts:

Yields 10 Tests/reconstituted vial, 100 Tests/kit.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 10231TM*
Description Candida albicans ATCC™ 10231™
Format Quanti-Cult Plus
Product Type Quality Control Organism
Target Organism Class Fungi
Test Type Quantitative QC
Packaging Type 100 Test/Kit
Quantity 100 Tests/Kit
Strain Designations 3149 [CBS 6431, CCY 29-3-106, CIP 48.72, DSM 1386, IFO 1594, NCPF 3179, NCYC 1363, NIH 3147, VTT C-85161]

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.