Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Quanti-Cult Plus™ Salmonella sp. serovar Abony NCTC 6017

Catalog No. R4716007 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4716007 100 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4716007 Supplier Thermo Scientific™ Supplier No. R4716007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.

Thermo Scientific™ Quanti-Cult Plus is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • Ready for rehydration
  • For use in quality control procedures

Includes:

10 preserved organism vials and 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® NCTC 6017
Description Salmonella sp. serovar Abony NCTC 6017
Format Quanti-Cult Plus
Product Type Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Quantitative QC
Packaging Type 100 Test/Kit
Quantity 100 Tests/Kit
Strain Designations CCM 4518 [NBRC 100797, NCTC 6017, CIP 80.39]

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.