Learn More
Thermo Scientific™ Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Quanti-Cult Plus™ Serratia marcescens ATCC™ 8100™ is supplied as a film of preserved microorganisms inside the cap on a plastic vial.
- No serial dilution required
- Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
- 10 tests/reconstituted vial
- Ready for rehydration
- For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
Specifications
Specifications
| ATCC® | 8100TM* |
| Description | Serratia marcescens ATCC™ 8100™ |
| Format | Quanti-Cult Plus |
| Product Type | Quality Control Kit |
| Target Organism Class | Gram-negative Rods |
| Test Type | Quantitative QC |
| Strain Designations | NRS 116-175 [CIP 104221] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.