Learn More
Thermo Scientific™ Quanti-Cult™ Pseudomonas aeruginosa ATCC™ 27853™
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Quanti-Cult™ Pseudomonas aeruginosa ATCC™ 27853™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial, ready for rehydration and use in quality control procedures.
- No serial dilution required
- Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum
Ordering Alerts:
Yield is 1 Test/reconstituted vial, 10 Tests/kit
Includes:
10 preserved organism vials and 10 rehydration vials.
Specifications
Specifications
| ATCC® | 27853TM* |
| Description | Pseudomonas aeruginosa ATCC™ 27853™ |
| Format | Quanti-Cult |
| Product Type | Bactidisk |
| Target Organism Class | Gram-negative Rods |
| Test Type | Quantitative QC |
| Packaging Type | 10 Test/Kit |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.