Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Quanti-Cult™ Staphylococcus aureus subsp. aureus ATCC™ 25923™

Catalog No. r4737010
Change view
Click to view available options
Quantity:
10 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4737010 10 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4737010 Supplier Thermo Scientific™ Supplier No. R4737010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media that can be stored at room-temperature to free up incubator space.

  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum

Ordering Alerts:

Yiels 1 Test/reconstituted vial, 10 Tests/kit

Includes:

10 preserved organism vials and 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 25923TM*
Description Staphylococcus aureus subsp. aureus ATCC™ 25923™
Format Quanti-Cult
Product Type Bactidisk
Target Organism Class Cocci
Test Type Quantitative QC
Packaging Type 10 Test/Kit
Quantity 10 Tests/Kit
Strain Designations Seattle 1944

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.