Learn More
Invivogen QUANTI-Luc™ 4 Lucia/Gaussia, Bioluminescent Assay Reagent, Real-Time Monitoring, Liquid
SDP
Catalog No. NC2251341
Encompass
QUANTI-Luc™ 4 Lucia/Gaussia is a bioluminescent assay reagent designed for precise quantitative detection of secreted luciferases. Its optimized formulation minimizes auto-luminescence and enhances detection sensitivity and range, making it ideal for gene expression and protein secretion studies. The dual luminescence modes offer versatility for quick end-point detection or high-throughput screening, allowing for dynamic real-time cellular monitoring with minimal sample disruption.
This item is not returnable. View return policy
Catalog No. NC2251341 Supplier Invivogen Supplier No. REPQLC4LG1
QUANTI-Luc™ 4 Lucia/Gaussia is a bioluminescent assay reagent designed for precise quantitative detection of secreted luciferases. Its optimized formulation minimizes auto-luminescence and enhances detection sensitivity and range, making it ideal for gene expression and protein secretion studies. The dual luminescence modes offer versatility for quick end-point detection or high-throughput screening, allowing for dynamic real-time cellular monitoring with minimal sample disruption.