Learn More
Invivogen QUANTI-Luc™ Coelenterazine-utilizing Luciferase Detection Medium
SDP
Catalog No. NC0762977
Encompass
QUaNTI-Luc - a coelenterazine-based luminescence assay reagent. Unit Size: 5 pouches (5 x 25ml). QUaNTI-Luc, is an assay reagent containing all the components required to quantitatively measure the activity of Lucia luciferase and other coelenterazine-utilizing luciferases. One pouch prepares 5 x 96 well plates.
Catalog No. NC0762977 Supplier Invivogen Supplier No. REPQLC2
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
QUaNTI-Luc - a coelenterazine-based luminescence assay reagent. Unit Size: 5 pouches (5 x 25ml). QUaNTI-Luc, is an assay reagent containing all the components required to quantitatively measure the activity of Lucia luciferase and other coelenterazine-utilizing luciferases. One pouch prepares 5 x 96 well plates.