Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Quantitative Multiplex Reference Standard (FFPE)
Catalog No. NC2739743
Encompass
The Quantitative Multiplex FFPE Reference Standard is a highly-characterized, biologically-relevant quality control material used to assess the performance of NGS assays that detect somatic mutations.
Catalog No. NC2739743 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. HD200
The Quantitative Multiplex FFPE Reference Standard is a highly-characterized, biologically-relevant quality control material used to assess the performance of NGS assays that detect somatic mutations.