Learn More
IDEXX LPD QUANTI-TRAY 97-WELL, 100 pack, Disposable, 100 mL sample volume, Sterile, Designed for bacterial enumeration using MPN method
Catalog No. NC1971767
Encompass
The IDEXX Quanti-Tray/2000 97-Well (100 pack) is a disposable tray designed for the rapid and reliable enumeration of bacteria in water and environmental samples using the Most Probable Number (MPN) method. This system minimizes hands-on time and ensures efficient microbial analysis, delivering high-confidence results typically within 24 hours.
  • Streamlines microbial quantification and reduces potential errors.
  • Superior 95% confidence limit compared to conventional methods.
  • Processes up to four samples per minute.
  • Requires less than 1 minute of hands-on time.
  • Features a sterile, disposable design.
  • Counting range of up to 2,419 MPN per 100 mL.
  • Includes 100 trays per pack.
  • Utilizes semiautomated processes for enhanced throughput.
  • Detection limit of 1 organism per 100 mL.
  • Shelf life of 18 months from the date of manufacture.
Catalog No. NC1971767 Supplier IDEXX LPD Supplier No. 982167500
The IDEXX Quanti-Tray/2000 97-Well (100 pack) is a disposable tray designed for the rapid and reliable enumeration of bacteria in water and environmental samples using the Most Probable Number (MPN) method. This system minimizes hands-on time and ensures efficient microbial analysis, delivering high-confidence results typically within 24 hours.
  • Streamlines microbial quantification and reduces potential errors.
  • Superior 95% confidence limit compared to conventional methods.
  • Processes up to four samples per minute.
  • Requires less than 1 minute of hands-on time.
  • Features a sterile, disposable design.
  • Counting range of up to 2,419 MPN per 100 mL.
  • Includes 100 trays per pack.
  • Utilizes semiautomated processes for enhanced throughput.
  • Detection limit of 1 organism per 100 mL.
  • Shelf life of 18 months from the date of manufacture.