Learn More
Electron Microscopy Sciences Perforated support foil with pre-defined holes has advantages in electron microscopy compared with conventional holey film. ~1.2 µm holes with 2.5 µm period. Magnifications between 30,000x and 40,000x.
SDP
Catalog No. NC2014604
Encompass_Preferred
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement.
Catalog No. NC2014604 Supplier Electron Microscopy Sciences Supplier No. Q350AR1.3
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement.