Learn More
Electron Microscopy Sciences Perforated support foil with pre-defined holes has advantages in electron microscopy compared with conventional holey film. 1 µm holes with 5 µm period. Magnifications between 30,000x and 40,000x.
SDP
Catalog No. NC3760731
Encompass
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. May be preferred over R 1.2/1.3, when an increased tolerance with respect to the position of beam, and a larger beam diameter are desired, such as in the case of automated image aquisition. Hole size is 1 µm. Space between holes is 3 µm. Center to center is 4 µm. Smaller holes are designed for magnifications of 50,000x or higher.
Catalog No. NC3760731 Supplier Electron Microscopy Sciences Supplier No. Q220CR14
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. May be preferred over R 1.2/1.3, when an increased tolerance with respect to the position of beam, and a larger beam diameter are desired, such as in the case of automated image aquisition. Hole size is 1 µm. Space between holes is 3 µm. Center to center is 4 µm. Smaller holes are designed for magnifications of 50,000x or higher.